MilliporeSigma
HLA dosage effect in narcolepsy with cataplexy.
Immunogenetics (2014-10-04)
Astrid van der Heide, Willem Verduijn, Geert W Haasnoot, Jos J M Drabbels, Gert J Lammers, Frans H J Claas
RÉSUMÉ
Narcolepsy with cataplexy is a sleep disorder caused by the loss of hypocretin-producing neurons in the hypothalamus. It is tightly associated with a specific human leukocyte antigen (HLA)-allele: HLA-DQB1*06:02. Based on this, an autoimmune process has been hypothesized. A functional HLA-DQ molecule consists of a DQα and a DQβ chain. HLA-DQB1*06:02 (DQβ) has a strong preference for binding to HLA-DQA1*01:02 (DQα), and together they form the functional DQ0602 dimer. A dosage effect would be expected if the HLA-DQ0602 dimer itself is directly involved in the aetiology. An increased expression of the HLA-DQ0602 dimer is expected in individuals homozygous for HLA-DQB1*06:02-DQA1*01:02, but is also hypothesized in individuals heterozygous for HLA-DQB1*06:02 and homozygous for HLA-DQA1*01:02. To study the impact of the expression of the HLA-DQ0602 dimer on narcolepsy susceptibility, 248 Dutch narcolepsy patients and 1272 Dutch control subjects, all of them positive for DQB1*06:02 (heterozygous and homozygous), were HLA-genotyped with attention not only to DQB1 but also to DQA1*01:02. DQB1*06:02-DQA1*01:02 homozygosity was significantly more often seen in patients compared to controls (O.R. 2.29) confirming previous observations. More importantly, a significantly higher prevalence of homozygosity for DQA1*01:02 was found in HLA-DQB1*06:02 heterozygous patients compared to controls (O.R. 2.37, p < 0.001). The latter finding clearly supports a direct role of the HLA-DQ molecule in the development of disease.