MilliporeSigma
Identification of a novel gene (ADPRTL1) encoding a potential Poly(ADP-ribosyl)transferase protein.
Genomics (2000-01-25)
I H Still, P Vince, J K Cowell
RÉSUMÉ
Poly(ADP-ribosyl)ation of nuclear proteins plays a significant role in the maintenance of genomic DNA stability. To date, four poly(ADP-ribosyl)ating proteins have been identified in humans. We now report the full-length sequence, expression profile, and chromosomal localization of a novel gene, ADPRTL1, encoding an ADP-ribosyltransferase-like protein. The predicted open reading frame encodes a protein of 1724 amino acids with a molecular mass of 192.8 kDa. The protein contains a region showing homology to the catalytic domains of the nuclear-localized ADP-ribosyltransferase proteins (Adprt), two recently identified Adprt-like proteins (Adprtl2 and Adprtl3), and the telomere-associated protein tankyrase. Key amino acids known to be important for the activity of these enzymes are conserved within this region of the Adprtl1 protein, indicating that Adprtl1 is a functional poly(ADP-ribosyl)transferase. As has been noted for tankyrase, sequence analysis of the Adprtl1 protein suggests that it is not capable of binding DNA directly. Thus, the transferase activity of Adprtl1 may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus. We have subsequently refined the location of the ADPRTL1 genomic locus to 13q11, close to the recently cloned ZNF198 gene.