MilliporeSigma
  • B4GALT1-congenital disorders of glycosylation presents as a non-neurologic glycosylation disorder with hepatointestinal involvement.
B4GALT1-congenital disorders of glycosylation presents as a non-neurologic glycosylation disorder with hepatointestinal involvement.
The Journal of pediatrics (2011-09-17)
Maïlys Guillard, Eva Morava, Jorg de Ruijter, Tony Roscioli, Johann Penzien, Lambert van den Heuvel, Michel A Willemsen, Arjan de Brouwer, Olaf A Bodamer, Ron A Wevers, Dirk J Lefeber
RÉSUMÉ
The clinical phenotype of congenital disorders of glycosylation is heterogeneous, mostly including a severe neurological involvement and multisystem disease. We identified a novel patient with a galactosyltransferase deficiency with mild hepatopathy and coagulation anomalies, but normal psychomotor development. The tissue-specific expression of the defective B4GALT1 gene correlated with the clinical phenotype.