MilliporeSigma
  • Recent natural selection identifies a genetic variant in a regulatory subunit of protein phosphatase 2A that associates with altered cancer risk and survival.
Recent natural selection identifies a genetic variant in a regulatory subunit of protein phosphatase 2A that associates with altered cancer risk and survival.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-09-24)
Lukasz F Grochola, Alexei Vazquez, Elisabeth E Bond, Peter Würl, Helge Taubert, Thomas H Müller, Arnold J Levine, Gareth L Bond
RÉSUMÉ
A regulated p53-dependent stress response is crucial in suppressing tumor formation and mediating the response to commonly used cancer therapeutics. However, little is known about the human, inherited genetics of this important signaling pathway. Studies of human genetic variants in the p53 tumor suppressor gene and MDM2 oncogene have shown that single nucleotide polymorphisms (SNP) can affect p53 signaling, confer cancer risk, and alter outcome, and also suggest that the pathway is under evolutionary selective pressure. Here, we attempt to accelerate the identification of functional p53 pathway SNPs by incorporating these characteristics into an analysis of 142 genes that are known to affect p53 signaling. We report that a genomic scan for recent natural selection denotes that of the 142 genes studied, the PPP2R5E gene that encodes a regulatory subunit of the tumor suppressing protein phosphatase 2A resides in a naturally selected genomic region. We go on to show that a selected SNP in PPP2R5E (epsilon-SNP2) associates with significant allelic differences in the onset (up to 19.2 years; P = 0.0002) and risk (odds ratio, up to 8.1; P = 0.0009) of soft tissue sarcoma development, as well as overall survival (relative risk, up to 3.04; P = 0.026). The PPP2R5E gene is identified as harboring genetic variants that can affect human cancer and are possibly under evolutionary selection pressure.