MilliporeSigma
  • Dialkyl phosphates in amniotic fluid as a biomarker of fetal exposure to organophosphates in Crete, Greece; association with fetal growth.
Dialkyl phosphates in amniotic fluid as a biomarker of fetal exposure to organophosphates in Crete, Greece; association with fetal growth.
Reproductive toxicology (Elmsford, N.Y.) (2014-04-02)
D Koutroulakis, S Sifakis, M N Tzatzarakis, A K Alegakis, E Theodoropoulou, M P Kavvalakis, D Kappou, A M Tsatsakis
RÉSUMÉ
The aim of this study was to evaluate fetal exposure to organophosphate pesticides (OPs) by measuring their non-specific dialkyl-phosphate metabolites (DAPs) in amniotic fluid (AF), and to examine the potential association between prenatal exposure and fetal growth. AF samples were collected from 415 women during the second gestational trimester. The determined OPs metabolites were DMP, DMTP, DEP, DETP, and DEDTP. DAPs were extracted by liquid-solid extraction, derivatized and analyzed by gas chromatography-mass spectrometry. 97.8% of AF samples were positive for at least one DAP. DAPs levels did not differ between urban and rural areas. Macrosomic neonates have significantly higher sum levels of DMPs (p=0.043), which exerted a linear positive association with birth-weight centile (b=4.43, p=0.016). Conclusively, as DAPs are detectable in AF they may be used as a potential biomarker of fetal exposure to OPs. Sum levels of DMPs appear to be associated with birth weight independently of other covariates.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
O,O-Diethyl thiophosphate potassium salt, 98%