- Identification of nonpeptidic small-molecule inhibitors of interleukin-2.
Identification of nonpeptidic small-molecule inhibitors of interleukin-2.
Bioorganic & medicinal chemistry letters (2005-02-03)
Nathan D Waal, Wenjin Yang, Johan D Oslob, Michelle R Arkin, Jennifer Hyde, Wanli Lu, Robert S McDowell, Chul H Yu, Brian C Raimundo
PMID15686897
RÉSUMÉ
The identification, design, and synthesis of a series of novel sulfamide- and urea-based small-molecule antagonists of the protein-protein interaction IL-2/IL-2Ralpha are described. Installation of a furan carboxylic acid fragment onto a low-micromolar sulfamide resulted in a 23-fold improvement in activity, providing a sub-micromolar, nonpeptidic IL-2 inhibitor (IC(50)=0.60 microM).