MilliporeSigma
  • Fas/FasL and perforin/granzyme pathway in acute rejection and diffuse alveolar damage after allogeneic lung transplantation-a human biopsy study.
Fas/FasL and perforin/granzyme pathway in acute rejection and diffuse alveolar damage after allogeneic lung transplantation-a human biopsy study.
Virchows Archiv : an international journal of pathology (2004-10-14)
Iris Bittmann, Christian Müller, Jürgen Behr, Jan Groetzner, Lorenz Frey, Udo Löhrs
RÉSUMÉ
Acute rejection and diffuse alveolar damage are major problems during the early time after transplantation. Against this background, lung biopsies after allogeneic lung transplantation were studied using immunohistochemistry. Biopsies with acute rejection, diffuse alveolar damage and morphological inconspicuous biopsies were chosen. The objectives of this study were to ascertain: (a) if and how CD4 and CD8 T cells contribute to allograft rejection and diffuse alveolar damage, (b) whether there is a correlation of the chemoattractant regulated on activation normal T cells (RANTES) with the mononuclear infiltrate and (c) whether perforin/granzyme and Fas/FasL pathways contribute to lung injury after lung transplantation. Our results show that CD4(+) and CD8(+) T cells were increased in biopsies with acute rejection and, to a minor extent, also in biopsies with diffuse alveolar damage due to reperfusion injury. RANTES expression of T cells was increased in biopsies with acute rejection. Perforin seemed to have a dual role in the alloimmune response. In one regard, it had a cytolytic function in the acute rejection process, and, in contrast, it may be responsible for downregulating both CD4- and CD8-mediated alloimmune responses. The FasL/Fas pathway is not only important for induction of apoptosis during rejection but is also a mechanism of lung injury in the development of diffuse alveolar damage.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Perforin (MRQ-23) Mouse Monoclonal Antibody