MilliporeSigma
E-Factor minimized hydrophosphonylation of aldehydes catalyzed by polystyryl-BEMP under solvent-free conditions.
Organic & biomolecular chemistry (2013-06-28)
Tommaso Angelini, Simona Bonollo, Daniela Lanari, Ferdinando Pizzo, Luigi Vaccaro
RÉSUMÉ
An efficient protocol for the hydrophosphonylation of aromatic and aliphatic aldehydes catalyzed by PS-BEMP under solvent-free conditions (SolFC) has been reported. Addition reactions were performed by using equimolar amounts of reagents and the resulting α-hydroxyphosphonates were isolated with simple workup procedures. A large scale protocol for the preparation of a representative α-hydroxyphosphonate 3a has been also set up using a flow reactor.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, purum, ≥98.0% (GC)