MilliporeSigma
Identification of alternatively spliced isoforms of interleukin-2/15 receptor β chain in ducks.
Veterinary immunology and immunopathology (2014-12-03)
Jipseol Jeong, Woo H Kim, Jaeseung Yeo, Cherry P Fernandez, Suk Kim, Youn-Jeong Lee, Hyun S Lillehoj, Wongi Min
RÉSUMÉ
Interleukin (IL)-2 and IL-15 receptor β (IL-2/15Rβ, CD122) play important roles in signal transduction for biological functions of IL-2 and IL-15. We found that ducks possess three different IL-2/15Rβ transcripts, a conventional form (duIL-2/15Rβ) and two variants. Comparisons between the cDNA and genomic sequences revealed that the two variants, duIL-2/15Rβ-d7 and duIL-2/15Rβ-d9, were novel spliced transcripts resulting from skipping exons 7 and 9, respectively. Expression profiles of duIL-2/15Rβ and its isoforms were examined in healthy tissues, concanavalin A (ConA)-stimulated splenic lymphocytes and in livers and spleens of Riemerella anatipestifer-infected ducks using quantitative real-time PCR (qRT-PCR). Generally, duIL-2/15Rβ-d9 expression was undetectable in healthy tissues, ConA-activated samples, and R. anatipestifer-infected ducks. Expression levels of duIL-2/15Rβ transcript were relatively high to moderate in all healthy tissues tested, while duIL-2/15Rβ-d7 expression was low. Compared to untreated controls, expression levels of duIL-2/15Rβ were elevated in ConA-activated splenic lymphocytes and in livers on day 7 in R. anatipestifer-infected ducks, while duIL-2/15Rβ-d7 expression was unchanged. Additionally, COS-7 cells transfected with duIL-2/15Rβ, duIL-2/15Rβ-d7, or duIL-2/15Rβ-d9 constructs generated 73 kilodalton (kDa), 31kDa, and 40kDa proteins, respectively. This study identified three different IL-2/15Rβ transcripts, including two isoforms generated by alternative splicing and their gene expression patterns in stimulated conditions.