MilliporeSigma
Mitochondrial cristae revealed with focused light.
Nano letters (2009-05-23)
Roman Schmidt, Christian A Wurm, Annedore Punge, Alexander Egner, Stefan Jakobs, Stefan W Hell
RÉSUMÉ
Because of the diffraction resolution barrier, optical microscopes have so far failed in visualizing the mitochondrial cristae, that is, the folds of the inner membrane of this 200 to 400 nm diameter sized tubular organelle. Realizing a approximately 30 nm isotropic subdiffraction resolution in isoSTED fluorescence nanoscopy, we have visualized these essential structures in the mitochondria of intact cells. We find a pronounced heterogeneity in the cristae arrangements even within individual mitochondrial tubules.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Abberior® STAR RED NHS, ≥80.0% (degree of coupling)