MilliporeSigma
Efficacy and safety of felodipine, a new dihydropyridine calcium channel blocker, in elderly hypertensive patients.
Clinical and experimental hypertension. Part A, Theory and practice (1992-01-01)
V DeQuattro
RÉSUMÉ
Felodipine is a new dihydropyridine calcium channel blocker with a number of properties that enhance its suitability as a first-line antihypertensive drug for the elderly. Felodipine has a 100-fold selectivity for inhibiting the contribution of vascular smooth muscle compared with cardiac muscle. Negative inotropic action appears minimal while selectivity appears to increase with age. Felodipine has a minimal effect on smooth muscle of venous capacitance vessels, thereby greatly reducing the likelihood of orthostatic hypotension. Renal effects are favorable; the glomerular filtration rate is increased in some patients. The extended-release formulation of felodipine produces a smooth 24-hour plasma concentration curve and is effective when prescribed once daily. Felodipine appears to lower blood pressure effectively in the elderly patient with few, generally mild, adverse effects.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Felodipine, solid
Felodipine, European Pharmacopoeia (EP) Reference Standard