MilliporeSigma
  • Structure of a 14-3-3 coordinated hexamer of the plant plasma membrane H+ -ATPase by combining X-ray crystallography and electron cryomicroscopy.
Structure of a 14-3-3 coordinated hexamer of the plant plasma membrane H+ -ATPase by combining X-ray crystallography and electron cryomicroscopy.
Molecular cell (2007-02-10)
Christian Ottmann, Sergio Marco, Nina Jaspert, Caroline Marcon, Nicolas Schauer, Michael Weyand, Caroline Vandermeeren, Geoffrey Duby, Marc Boutry, Alfred Wittinghofer, Jean-Louis Rigaud, Claudia Oecking
RÉSUMÉ
Regulatory 14-3-3 proteins activate the plant plasma membrane H(+)-ATPase by binding to its C-terminal autoinhibitory domain. This interaction requires phosphorylation of a C-terminal, mode III, recognition motif as well as an adjacent span of approximately 50 amino acids. Here we report the X-ray crystal structure of 14-3-3 in complex with the entire binding motif, revealing a previously unidentified mode of interaction. A 14-3-3 dimer simultaneously binds two H(+)-ATPase peptides, each of which forms a loop within the typical 14-3-3 binding groove and therefore exits from the center of the dimer. Several H(+)-ATPase mutants support this structure determination. Accordingly, 14-3-3 binding could result in H(+)-ATPase oligomerization. Indeed, by using single-particle electron cryomicroscopy, the 3D reconstruction of the purified H(+)-ATPase/14-3-3 complex demonstrates a hexameric arrangement. Fitting of 14-3-3 and H(+)-ATPase atomic structures into the 3D reconstruction map suggests the spatial arrangement of the holocomplex.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Fusicoccin from Fusicoccum amygdali, ≥85% (HPLC)