MilliporeSigma
Synthesis of Ag2S quantum dots in water-in-CO2 microemulsions.
Chemical communications (Cambridge, England) (2004-11-16)
Juncheng Liu, Poovathinthodiyil Raveendran, Zameer Shervani, Yutaka Ikushima
RÉSUMÉ
Ag2S nanocrystals with a mean diameter of 5.9 nm (sigma= 1.65 nm) and characteristic surface plasmon resonance absorption at 330 nm have been synthesized in water-in-supercritical CO2 reverse microemulsion using the commonly used AOT surfactant with 2,2,3,3,4,4,5,5-octafluoro-1-pentanol (F-pentanol) as cosurfactant.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%