MilliporeSigma
Synthesis of a leucomitosane via a diastereoselective radical cascade.
The Journal of organic chemistry (2013-06-01)
François Brucelle, Philippe Renaud
RÉSUMÉ
The preparation of trans-2,3-disubstituted indolines from 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade using ethyl iodoacetate and triethylborane is described. Further lactamization afforded substituted benzopyrrolizidinones with excellent diastereomeric ratios. The radical cascade/lactamization sequence was efficiently applied to the synthesis of a 3-oxo-leucomitosane related to the mitomycin family of alkaloids.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Ethyl iodoacetate, 98%