MilliporeSigma
[Studies of the chemical constituents of Ardisia pusilla A. DC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1990-03-01)
X Wang, Q Zhang
RÉSUMÉ
Six chemical compounds isolated from the leaves and stems of Ardisia pusilla were identified as gallic acid (I), succinic acid (II), naringenin-6-C-glucoside (III), kaempferol-3-O-beta-D-galactoside (IV) by chemical and spectroscopic analysis. They were isolated from Ardisia plant for the first time.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Naringenin-6-C-glucoside, ≥95% (LC/MS-ELSD)