MilliporeSigma
Pd-catalyzed C3-selective arylation of pyridines with phenyl tosylates.
Chemical communications (Cambridge, England) (2013-04-12)
Fenglin Dai, Qingwen Gui, Jidan Liu, Zhiyong Yang, Xiang Chen, Ruqing Guo, Ze Tan
RÉSUMÉ
We have discovered that phenyl tosylates can be used to arylate pyridines at the C3-position using a Pd(OAc)2-1,10-phenanthroline catalyst system. We also discovered that the reaction of 4-methylpyridine with naphthyl tosylates occurred on the methyl group instead of at the C3-position.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Methylpyridine, 99%