MilliporeSigma
Role of STRAP in regulating GSK3β function and Notch3 stabilization.
Cell cycle (Georgetown, Tex.) (2011-04-20)
Nilesh D Kashikar, Wanguang Zhang, Pierre P Massion, Adriana L Gonzalez, Pran K Datta
RÉSUMÉ
Glycogen synthase kinase 3β (GSK3β) can regulate a broad range of cellular processes in a variety of cell types and tissues through its ability to phosphorylate its substrates in a cell- and time-specific manner. Although it is known that Axin and presenilin help to recruit β-catenin/Smad3 and tau protein to GSK3β, respectively, it is not clear how many of the other GSK3β substrates are recruited to it. Here, we have established the binding of GSK3β with a novel scaffold protein, STRAP, through its WD40 domains. In a new finding, we have observed that STRAP, GSK3β and Axin form a ternary complex together. We show for the first time that intracellular fragment of Notch3 (ICN3) binds with GSK3β through the ankyrin repeat domain. This binding between STRAP and GSK3β is reduced by small-molecule inhibitors of GSK3β. Further studies revealed that STRAP also binds ICN3 through the ankyrin repeat region, and this binding is enhanced in a proteasomal inhibition-dependent manner. In vivo ubiquitination studies indicate that STRAP reduces ubiquitination of ICN3, suggesting a role of STRAP in stabilizing ICN3. This is supported by the fact that STRAP and Notch3 are co-upregulated and co-localized in 59% of non-small cell lung cancers, as observed in an immunohistochemical staining of tissue microarrays. These results provide a potential mechanism by which STRAP regulates GSK3β function and Notch3 stabilization and further support the oncogenic functions of STRAP.