MilliporeSigma
Convenient ultrasound-mediated synthesis of 1,4-diazabutadienes under solvent-free conditions.
Ultrasonics sonochemistry (2010-08-28)
Jing-Yu He, Hong-Xing Xin, Hong Yan, Xiu-Qing Song, Ru-Gang Zhong
RÉSUMÉ
An ultrasound-assisted preparation of 1,4-diazabutadienes via smooth condensation of diketones with amines under solvent-free conditions is described. The generality of this method was examined by the synthesized N,N'-diaryl- and N,N'-dialkyl-1,4-diazabutadiene derivatives. In addition to experimental simplicity, the main advantages of the procedure are mild conditions, short reaction time (2-15 min) and high yields (71-98%).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,5-Difluoroaniline, 99.5%