MilliporeSigma
Determination of linsidomine in human plasma by tandem LC-MS with ESI.
Journal of pharmaceutical and biomedical analysis (2000-04-15)
F C Sutherland, A D de Jager, K J Swart, H K Hundt, T Scanes, A F Hundt
RÉSUMÉ
A sensitive method for the determination of linsidomine in plasma was developed, using high-performance liquid chromatographic (HPLC) separation with tandem mass spectrometric detection. Linsidomine was derivatised with propyl chloroformate and extracted with tert-butyl methyl ether/1,2-dichloroethane (55:45, v/v), back-extracted into HCl (0.01 M) followed by alkalinisation and back-extraction into ether; the final ether extract evaporated, reconstituted in mobile phase and then separated on a Phenomenex Luna C18 (2) 5 micron 2.1 x 150 mm column with a mobile phase consisting of methanol water formic acid (98/100%) (400:600:0.05, v/v/v) at a flow-rate of 0.4 ml min(-1). Detection was achieved by a Finnigan MAT mass spectrometer (LCQ) at unit resolution in the selected reaction monitoring (SRM) mode monitoring the transition of the protonated molecular ion m/z 257.0 to the product ion m/z 86.0. The mean recovery for linsidomine was 51% with a lower limit of quantification of 0.70 ng/ml using 1 ml plasma for extraction. This LC-MS/MS method for the determination of linsidomine in human plasma allows for better specificity and a higher sample throughput than the traditional LC-UV methods. It also demonstrates the profound effect that the composition of acidic modifiers and matrix constituents can have on the electrospray ionisation (ESI) of the analyte.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Butyl chloroformate, 98%