MilliporeSigma
  • Matrix-assisted ultraviolet laser-desorption ionization time-of-flight mass spectrometry of sulfated mannans from the red seaweed Nothogenia fastigiata.
Matrix-assisted ultraviolet laser-desorption ionization time-of-flight mass spectrometry of sulfated mannans from the red seaweed Nothogenia fastigiata.
Carbohydrate research (2000-11-22)
R Erra-Balsells, A A Kolender, M C Matulewicz, H Nonami, A S Cerezo
RÉSUMÉ
Matrix-assisted ultraviolet laser-desorption ionization time-of-flight mass spectrometry (UV-MALDI-TOF-MS) was applied to sulfated xylo-mannan fractions from Nothogenia fastigiata in order to determine their molecular weights and distribution profiles. The number-average molecular weight calculated from the spectra was similar to that determined by chemical end-group analysis for the lower molecular weight fractions. For the other fractions, the number-average molecular weight was lower than that chemically determined; the increased difference may be attributed to higher desorption difficulties and, consequently, mass-dependent discrimination. A reconstructed spectrum, using the peaks obtained from all the fractions, suggested an unimodal distribution. The best results were obtained by using 2,5-dihydroxybenzoic acid as matrix doped with 1-hydroxyisoquinoline and with harmane and nor-harmane.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Isocarbostyril, 98%