MilliporeSigma
Proteomic analysis of oropharyngeal carcinomas reveals novel HPV-associated biological pathways.
International journal of cancer (2012-06-27)
Robbert J C Slebos, Nico Jehmlich, Brandee Brown, Zhirong Yin, Christine H Chung, Wendell G Yarbrough, Daniel C Liebler
RÉSUMÉ
Oropharyngeal carcinoma (OPC) can be classified into two equally prevalent subtypes depending on the presence of human papillomavirus (HPV). Patients with HPV-positive (HPV+) OPC represent a unique cohort with a distinct tumor biology and clinical behavior compared to HPV-negative (HPV-) OPC. Genetic studies have demonstrated chromosomal and gene expression changes associated with distinct subclasses of OPC; however, the proteomic consequences of HPV infection are not known. We analyzed sets of ten HPV+ and ten HPV- OPCs and ten normal adult oral epithelia using a standardized global proteomic analysis platform. This analysis yielded a total of 2,653 confidently identified proteins from which we chose 31 proteins on the basis of expression differences between HPV+, HPV- and normal epithelium for targeted protein quantitation. Analysis of differentially expressed proteins by HPV status revealed enrichment of proteins involved in epithelial cell development, keratinization and extracellular matrix organization in HPV- OPC, whereas enrichment of proteins in DNA initiation and replication and cell cycle control was found for HPV+ OPC. Enrichment analysis for transcription factor targets identified transcription factors E2F1 and E2F4 to be highly expressed in HPV+ OPC. We also found high expression of argininosuccinate synthase 1 in HPV+ OPC, suggesting that HPV+ OPC is more dependent on conditionally essential amino acid, arginine, and this was confirmed on a OPC-specific tissue microarray. These identified proteomic changes reveal novel driving molecular pathways for HPV+ and HPV- OPCs that may be pertinent in therapeutic strategies and outcomes of OPC.