MilliporeSigma
Synthesis and highly selective bromination of Azacalix[4]pyrimidine macrocycles.
The Journal of organic chemistry (2010-01-02)
Li-Xia Wang, De-Xian Wang, Zhi-Tang Huang, Mei-Xiang Wang
RÉSUMÉ
A number of N-substituted azacalix[4]pyrimidines were synthesized by two methods. While straightforward condensation reaction between 4,6-dichloropyrimidine and 4,6-bis(alkylamino)pyrimidines gave identically N-substituted azacalix[4]pyrimidines in low yields, a general and moderate-to-high yielding 1 + 3 macrocyclic fragment coupling reaction afforded azacalix[4]pyrimidines that contained either the same or different N-substituents. Upon treatment with N-bromosuccinimide (NBS) under controlled conditions, methylazacalix[4]pyrimidine was selectively brominated at lower rim to produce mono-, di-, and tribrominated azacalix[4]pyrimidines in good yields. While azacalix[4]pyrimidine derivatives adopted 1,3-alternate conformation in the solid state, the synthesized macrocycles were fluxional in solution, and the interconversion of various conformational structures was rapid relative to the NMR time scale.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4,6-Dichloropyrimidine, 97%