MilliporeSigma
The deoxyxylulose phosphate pathway of terpenoid biosynthesis in plants and microorganisms.
Chemistry & biology (1998-09-30)
W Eisenreich, M Schwarz, A Cartayrade, D Arigoni, M H Zenk, A Bacher
RÉSUMÉ
Recent studies have uncovered the existence of an alternative, non-mevalonate pathway for the formation of isopentenyl pyrophosphate and dimethylallyl pyrophosphate, the two building blocks of terpene biosynthesis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Deoxy-D-xylulose-5-phosphate sodium salt, ≥99.0% (TLC)