- The importance of GLU361 position in the reaction catalyzed by cholesterol oxidase.
The importance of GLU361 position in the reaction catalyzed by cholesterol oxidase.
Bioorganic & medicinal chemistry letters (1999-01-05)
I J Kass, N S Sampson
PMID9873599
RÉSUMÉ
Cholesterol oxidase stereospecifically isomerizes cholest-5-en-3-one to cholest-4-en-3-one. When the base catalyst for isomerization, Glu361, is mutated to Asp, the rate of deprotonation of cholest-5-en-3-one is not affected, but protonation of the dienolic intermediate becomes rate-limiting. This may be a consequence of the large distance between the catalytic base and carbon-6 of the intermediate in the mutant enzyme.
MATÉRIAUX