MilliporeSigma
Phase II clinical trial of a vas deferens injectable contraceptive for the male.
Contraception (1997-12-31)
S K Guha, G Singh, S Ansari, S Kumar, A Srivastava, V Koul, H C Das, R L Malhotra, S K Das
RÉSUMÉ
Following up on an earlier clinical trial demonstrating the safety of an intra-vas deferens injection of a contraceptive drug named Risug, comprised of styrene maleic anhydride (SMA) in a solvent vehicle of dimethylsulphoxide (DMSO), a study to assess the contraceptive effectiveness of a specific dose (60 mg) of SMA bilaterally was planned and implemented. Male subjects and their wives with normal reproductive profiles were the volunteer subjects. The wives were not using any contraceptives. The results reconfirm the safety and show that for a period of at least 1 year, the treatment leads to azoospermia in the male and gives pregnancy protection.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Poly(styrene-alt-maleic acid) sodium salt solution, 13 wt. % in H2O