MilliporeSigma
Quinazoline-2,4(1H,3H)-dione as a substitute for thymine in triple-helix forming oligonucleotides: a reassessment.
Nucleic acids research (1996-03-15)
J Michel, J J Toulmé, J Vercauteren, S Moreau
RÉSUMÉ
A major limitation in triple-helix formation arises from the weak energy of interaction between the third strand and the double-stranded target. We tried to increase the stacking interaction contribution within the third strand by extending the aromatic domain of thymine. We report here the use of 2,4-quinazolinedione as a substitute for thymine in the canonical TA*T triplet. The synthesis and the characterization of the quinazoline beta nucleoside Q and of its phosphoramidite derivative is described. Triple-helix- forming oligonucleotides incorporating Q have been prepared and their ability to form triplexes has been evaluated by UV-monitored thermal denaturation measurements. The introduction of one or multiple Q residues, either contiguous or remote from each other, slightly destabilized triple-stranded structures, whatever the nucleic acid base composition (pyrimidine or GT) of the third strand.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Benzoyleneurea, 97%