MilliporeSigma
1-(Tetrahydro-2-furanyl)-5-fluorouracil (Ftorafur) determined in rat hair as an index of drug exposure.
Journal of pharmaceutical sciences (1993-12-01)
T Uematsu, N Miyazawa, K Wada
RÉSUMÉ
The fluorescence derivatization of 1-(tetrahydro-2-furanyl)-5-fluorouracil (Ftorafur, FT) with 4-bromomethyl-6,7-dimethoxycoumarin (BrMdmc) with 18-crown-6 as catalyst was utilized for a sensitive and selective liquid chromatographic method to determine the concentration of FT in hair. Hair samples collected from rats, which had received FT intraperitoneally for 1 to 4 weeks, were dissolved in 1 M NaOH by heating at 80 degrees C for 30 min. A three-step extraction procedure for FT in the dissolved hair was used before and after the derivatization of FT with BrMdmc. The detection limit achieved was < 0.16 ng/mg hair. A dose-dependent accumulation of FT was evident in the rat hair (r = 0.914, p < 0.001): 0.24 +/- 0.07, 1.35 +/- 0.39, and 2.85 +/- 0.74 ng/mg hair (mean +/- SD, n = 4-5) at the doses of 5, 15, and 50 mg/kg/day for 4 weeks, respectively. In the rats that had received the highest dose of FT, the accumulation of FT in the hair depended also on the duration of FT administration: 0.82 +/- 0.33 (1 week), 1.52 +/- 0.38 (2 weeks), and 2.85 +/- 0.74 (4 weeks) ng/mg hair (n = 4-5). These findings suggest that the analysis of an oral antineoplasmic FT in the hair may be useful for assessing the degree of exposure to the drug.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Bromomethyl-6,7-dimethoxycoumarin, 96%