MilliporeSigma
  • Active-site peptides of acetylcholinesterase of Electrophorus electricus: labelling of His-440 by 1-bromo-[2-14C]pinacolone and Ser-200 by tritiated diisopropyl fluorophosphate.
Active-site peptides of acetylcholinesterase of Electrophorus electricus: labelling of His-440 by 1-bromo-[2-14C]pinacolone and Ser-200 by tritiated diisopropyl fluorophosphate.
Biochimica et biophysica acta (1994-10-19)
E Salih, S B Chishti, P Vicedomine, S G Cohen, D C Chiara, J B Cohen
RÉSUMÉ
To characterize the structure of the active site of acetylcholinesterase (AChE) from the electric organ of E. electricus, we identified sites of incorporation of two active-site affinity labels, [3H]diisopropyl fluorophosphate ([3H]DFP), and 1-bromo-2-[14C]pinacolone ([14C]BrPin). AChE was isolated, purified, inactivated and digested with trypsin, and peptides containing 3H or 14C were purified by reverse-phase HPLC and characterized by N-terminal sequence analysis. [3H]DFP, labelling Ser-200, was found in a single peptide, QVTIFGESAGAASVGMHLLSPDSR, 83% identical with the sequence from Thr-193 to Arg-216 deduced for AChE of T. californica, with Gln, Ala, Leu, and Asp in place of Thr-193, Gly-203, Ile-210 and Gly-214, respectively, and 87% identical with that from bovine and human brain AChEs. Inactivation by [14C]BrPin led to two radioactive peptides. One, ASNLVWPEWMGVIHGYEIEFVFGLPLEK, was 96% identical with that extending from Ala-427 to Lys-454 of T. californica. Release of 14C in cycle 14 established reaction of [14C]BrPin with active-site His-440, protected by 5-trimethylammonio-2-pentanone (TAP). The other peptide, LLXVTENIDDAER, 77% homologous with that of T. californica extending from Leu-531 to Arg-543, had label associated with the third cycle, not protected by TAP, corresponding to Asn-533. The slow inactivation of eel AChE by reaction of [14C]BrPin at His-440 contrasts with that of AChE from T. nobiliana, where it reacts rapidly with a free cysteine, Cys-231, not present in eel AChE. For both AChEs, inactivation by BrPin prevents subsequent reaction with [3H]DFP, and prior inactivation by DFP does not prevent reactions with [14C]BrPin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Bromopinacolone, ≥97%
Sigma-Aldrich
1-Bromopinacolone, technical grade, ~90%