MilliporeSigma
[Interaction of pipolfen and suprastin with human serum albumin depending on the pH of the medium and its calcium ion content].
Farmakologiia i toksikologiia (1983-11-01)
K I Bender, A N Lutsevich
RÉSUMÉ
Experiments made with the use of equilibrium dialysis and differential spectrophotometry have shown that pipolfen and difenhydramine interact actively with human serum albumin at pH 6.8-7.4. Calcium ions have been demonstrated to activate the interaction of pipolphen and diphenhydramine with albumin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Chloropyramine hydrochloride, analytical standard, for drug analysis