MilliporeSigma
Linking of N4-aminocytosine to glutathione by use of bromopyruvate.
Nucleic acids symposium series (1979-01-01)
N Nitta, K Sugihara, O Kuge, K Negishi, Y Wataya, H Hayatsu
PMID547239
RÉSUMÉ
N4-Aminocytosine reacted with acetone and acetaldehyde to form hydrazones that were readily revertible to the parent compound. With pyruvate, in contrast, it formed a stable hydrazone. By use of bromopyruvate, N4-aminocytosine was linked to glutathione.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Hydrazino-1H-pyrimidin-2-one, AldrichCPR