MilliporeSigma
Cytotoxicity and induction of repairable DNA damage by photoactivated 5-nitrofurfural.
Toxicology (1987-07-01)
R W Busker, G M Beijersbergen van Henegouwen
RÉSUMÉ
5-Nitrofurfural (NFA) an important photodecomposition product and metabolite of medicinal nitrofurans is phototoxic in bacterial test systems. Its major photodecomposition product 5-hydroxymethylene-2(5H)-furanone (HMF) appears to be responsible for this. Furthermore HMF and photoactivated nitrofurfural can induce repairable DNA damage in Escherichia coli bacteria. These effects may be important with respect to skin and allergic reactions and carcinogenicity appearing in nitrofuran therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Nitro-2-furaldehyde, 99%