MilliporeSigma
Inhibition of NADH oxidation by pyridine derivatives.
Biochemical and biophysical research communications (1987-07-15)
R R Ramsay, K A McKeown, E A Johnson, R G Booth, T P Singer
RÉSUMÉ
The neurotoxicity of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine, an impurity in an illicit drug, is expressed after its oxidation to 1-methyl-4-phenylpyridinium by monoamine oxidase. The pyridinium is concentrated by carrier-mediated transport into the mitochondria where it inhibits NADH dehydrogenase and, hence, ATP synthesis. Some structurally related compounds have been tested for their effect on the oxidation of NAD+-linked substrates in intact mitochondria, and for the inhibition of the accumulation of the pyridinium into mitochondria and of NADH dehydrogenase activity in a membrane preparation. Some pyridine derivatives are more inhibitory to NADH dehydrogenase than is 1-methyl-4-phenylpyridinium but these are not concentrated into mitochondria by the uptake system. 4-Phenylpyridine, one of the most effective inhibitors, both occurs naturally and is an environmental pollutant.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Phenylpyridine, 97%