- Integumentomuscular sac of Ascaris lumbricoides suis L: a biomembrane for studies on the transport of chemical substances.
Integumentomuscular sac of Ascaris lumbricoides suis L: a biomembrane for studies on the transport of chemical substances.
Polish journal of pharmacology and pharmacy (1986-07-01)
M Grzywacz
PMID2430266
RÉSUMÉ
The whole integumentomuscular sac of the worm Ascaris lumbricoides suis L. and pouches isolated from it seemed to be a good model for investigation of the transmembrane transport of chemical substances across tissue barriers.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Piperazine adipate, European Pharmacopoeia (EP) Reference Standard