MilliporeSigma
Pathological laughter.
Annals of emergency medicine (1990-03-01)
G L Zellers, M Frank, J Dougherty
RÉSUMÉ
A man experienced one hour and 40 minutes of continual, inappropriate, uncontrollable laughter. The onset was preceded by a single-inhalation exposure to an insecticide of very low toxicity. The episode was terminated by a single dose of IV diazepam. A discussion of pathological laughter, including its proposed pathophysiology, differential diagnosis, clinical relevance, and management, is presented.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
MGK 264, PESTANAL®, analytical standard