MilliporeSigma
  • Facile aqueous-phase synthesis of biocompatible and fluorescent Ag2S nanoclusters for bioimaging: tunable photoluminescence from red to near infrared.
Facile aqueous-phase synthesis of biocompatible and fluorescent Ag2S nanoclusters for bioimaging: tunable photoluminescence from red to near infrared.
Small (Weinheim an der Bergstrasse, Germany) (2012-07-11)
Chuanxi Wang, Yu Wang, Lin Xu, Dan Zhang, Mingxia Liu, Xiangwei Li, Hongchen Sun, Quan Lin, Bai Yang
RÉSUMÉ
Low toxicity and fluorescent nanomaterials have many advantages in biological imaging. Herein, a novel and facile aqueous-phase approach to prepare biocompatible and fluorescent Ag(2)S nanoclusters (NCs) is designed and investigated. The resultant Ag(2)S NCs show tunable luminescence from the visible red (624 nm) to the near infrared (NIR; 724 nm) corresponding to the increasing size of the NCs. The key for preparing tunable fluorescent Ag(2)S NCs is the proper choice of capping reagent, glutathione (GSH), and the novel sulfur-hydrazine hydrate complex as the S(2-) source. As a naturally occurring and readily available tripeptide, GSH functions as an important scaffold to prevent NCs from growing large nanoparticles. Additionally, GSH is a small biomolecule with several functional groups, including carboxyl and amino groups, which suggests the resultant Ag(2)S NCs are well-dispersed in aqueous solution. These advantages make the as-prepared Ag(2)S NCs potentially applicable to biological labeling as well. For example, the resultant Ag(2)S NCs are used as a probe for MC3T3-EI cellular imaging.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis