MilliporeSigma
Alcoholic hepatitis: a clinician's guide.
Clinics in liver disease (2012-05-01)
Gina Choi, Bruce Allen Runyon
RÉSUMÉ
Alcoholic hepatitis is a frequent reason for admission and a common consultation request for hepatologists and gastroenterologists. Although it seems to occur acutely, it is usually subacute and often superimposed on underlying alcoholic cirrhosis. Typically patients have a background of drinking on a daily basis, but, in response to a life crisis, patients have started drinking massively.