MilliporeSigma
Shiga toxin pathogenesis: kidney complications and renal failure.
Current topics in microbiology and immunology (2011-10-11)
Tom G Obrig, Diana Karpman
RÉSUMÉ
The kidneys are the major organs affected in diarrhea-associated hemolytic uremic syndrome (D(+)HUS). The pathophysiology of renal disease in D(+)HUS is largely the result of the interaction between bacterial virulence factors such as Shiga toxin and lipopolysaccharide and host cells in the kidney and in the blood circulation. This chapter describes in detail the current knowledge of how these bacterial toxins may lead to kidney disease and renal failure. The toxin receptors expressed by specific blood and resident renal cell types are also discussed as are the actions of the toxins on these cells.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Globotriaosylsphingosine from porcine blood