MilliporeSigma
Use of hydrogen as a carrier gas for the analysis of steroids with anabolic activity by gas chromatography-mass spectrometry.
Journal of chromatography. A (2011-09-13)
J A Muñoz-Guerra, P Prado, S Vargas García-Tenorio
RÉSUMÉ
Due to the impact in the media and the requirements of sensitivity and robustness, the detection of the misuse of forbidden substances in sports is a really challenging area for analytical chemistry, where any study focused on enhancing the performance of the analytical methods will be of great interest. The aim of the present study was to evaluate the usefulness of using hydrogen instead of helium as a carrier gas for the analysis of anabolic steroids by gas chromatography-mass spectrometry with electron ionization. There are several drawbacks related with the use of helium as a carrier gas: it is expensive, is a non-renewable resource, and has limited availability in many parts of the world. In contrast, hydrogen is readily available using a hydrogen generator or high-pressure bottled gas, and allows a faster analysis without loss of efficiency; nevertheless it should not be forgotten that due to its explosiveness hydrogen must be handled with caution. Throughout the study the impact of the change of the carrier gas will be evaluated in terms of: performance of the chromatographic system, saving of time and money, impact on the high vacuum in the analyzer, changes in the fragmentation behaviour of the analytes, and finally consequences for the limits of detection achieved with the method.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Epitestosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®