MilliporeSigma
  • Augmentation of immune response by chicoric acid through the modulation of CD28/CTLA-4 and Th1 pathway in chronically stressed mice.
Augmentation of immune response by chicoric acid through the modulation of CD28/CTLA-4 and Th1 pathway in chronically stressed mice.
Neuropharmacology (2011-01-29)
Kiranjeet Kour, Sarang Bani
RÉSUMÉ
This study demonstrates the protective effect of chicoric acid (CA) on chronic restraint stress-induced altered T lymphocyte subset distribution and corresponding cytokine secretion patterns in experimental Swiss albino mice. CA has the potential to restore diminished immune response and Th1/Th2 homeostasis in chronically stressed mice as evident by significant increase in lymphocyte proliferation and CD3(+), CD4(+) and CD8(+) T cell population. Interestingly, chicoric acid imparted immunostimulation mainly by upregulating the expression of CD28 and CD80 and downregulating CTLA-4. It exerted stimulatory effect on IL-12, IFN-gamma and IL-2 and suppressed the increased IL-10 levels in chronically stressed mice. It also exhibited a significant lowering effect on raised corticosterone levels and reversed the chronic stress-induced hypertrophy of adrenal glands and atrophy of thymus and spleen, thereby showing its normalizing effect on HPA axis. Our results reveal that CA has the potential to reverse the impact of chronic restraint stress on immune status by normalizing corticosterone levels and augmenting Th1 cytokine profile along with the co-stimulatory molecules particularly CD28/CTLA-4 pathway that plays a very important role in generation of an effective immune response in immune compromised situations.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
(−)-Chicoric acid, analytical standard