MilliporeSigma
Purification, characterization, and gene cloning of glucose-1-phosphatase from Citrobacter braakii.
The Journal of general and applied microbiology (2009-11-27)
Young-Ok Kim, Han-Woo Kim, In-Suk Park, Jeong-Ho Lee, Sang-Jun Lee, Kyung-Kil Kim
RÉSUMÉ
Citrobacter braakii produced an intracellular acid glucose phosphatase (AgpC) which was purified 986 fold to homogeneity with the specific activity of 286 units/mg. AgpC hydrolyzed a wide variety of phosphorylated compounds with high activity for glucose-1-phosphate and glucose-6-phosphate. The optimum pH and temperature for the enzyme activity was pH 5.0 and 45 degrees C, respectively. The Km value for glucose-1-phosphate was 5.12 mM with a Vmax 27.8 U mg(-1). Its molecular weight was 46 kDa by SDS-PAGE gel and the sequence of N-terminal amino acid residues identified was Gln-Thr-Ala-Pro-Glu-Gly-Tyr-Gln-Leu-Gln. The glucose-1-phosphatase gene (agpC) was cloned from the C. braakii genomic library. This gene comprised 1,242 nucleotides and encoded a polypeptide of 413 amino acids. The result of its BLAST search showed a significant similarity with glucose-1-phosphatase from enterobacteria such as E. coli, Enterobacter, Shigella, and Salmonella.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ala-Pro hydrate