MilliporeSigma
  • Tuning mechanism in a two-component columnar host system composed of 1,2-diphenylethylenediamine and 1,1'-binaphthyl-2,2'-dicarboxylic acid.
Tuning mechanism in a two-component columnar host system composed of 1,2-diphenylethylenediamine and 1,1'-binaphthyl-2,2'-dicarboxylic acid.
Organic letters (2008-01-11)
Yoshitane Imai, Kakuhiro Kawaguchi, Katuzo Murata, Tomohiro Sato, Reiko Kuroda, Yoshio Matsubara
RÉSUMÉ
In a two-component columnar host system composed of racemic (rac)-1,2-diphenylethylenediamine and rac-1,1'-binaphthyl-2,2'-dicarboxylic acid, a cavity tuning mechanism resulted from changes in the structure of the columns using a specific combination of the following four molecules: (1R,2R)-1, (1S,2S)-1, (R)-2, and (S)-2.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N,N′-Diphenylethylenediamine, 98%