MilliporeSigma
  • Determination of haloperidol and reduced haloperidol in human serum by liquid chromatography after fluorescence labeling based on the Suzuki coupling reaction.
Determination of haloperidol and reduced haloperidol in human serum by liquid chromatography after fluorescence labeling based on the Suzuki coupling reaction.
Analytical and bioanalytical chemistry (2006-09-08)
Naoya Kishikawa, Chiyuki Hamachi, Yoshihiro Imamura, Yoshihito Ohba, Kenichiro Nakashima, Yashuhiro Tagawa, Naotaka Kuroda
RÉSUMÉ
A simultaneous method for the determination of haloperidol (HP) and its metabolite, reduced haloperidol (RHP), in human serum was developed by means of high-performance liquid chromatography (HPLC) with fluorescence detection. Suzuki coupling reaction with a fluorescent arylboronic acid, 4-(4,5-diphenyl-1H-imidazol-2-yl)phenylboronic acid (DPA), was employed to convert HP and RHP into highly fluorescent compounds. HP and RHP were extracted from human serum by liquid-liquid extraction with a mixture of n-hexane and isoamyl alcohol (99:1, v/v) and subsequently labeled by reaction with DPA. Separation of DPA derivatives of HP and RHP was performed on a silica column with a mixture of acetonitrile and H(2)O (90:10, v/v) containing triethylamine and acetic acid as a mobile phase. The proposed method allowed sensitive detection of HP and RHP in human serum with a detection limit (at a signal to noise ratio of 3) of 0.22 and 0.20 ng/mL, respectively. The applicability of the method for therapeutic drug monitoring (TDM) was demonstrated by analyzing human serum samples from schizophrenic patients receiving HP.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Haloperidol metabolite II, analytical standard