MilliporeSigma
  • Detection of Nepsilon-monomethyllysine using high-performance liquid chromatography and high-performance liquid chromatography-mass spectrometry.
Detection of Nepsilon-monomethyllysine using high-performance liquid chromatography and high-performance liquid chromatography-mass spectrometry.
Journal of chromatography. A (2005-07-26)
H Kalász, Z Szücs, M Tihanyi, A Szilágyi, J Lengyel
RÉSUMÉ
Nepsilon-Monomethyllysine was identified in the serum, urine, brain, and liver samples of rats treated per os with L-deprenyl. The identification procedure included reaction with Fmoc chloride, clean-up, and analysis using HPLC-UV-MS. Oral administration of (-)-N-14C-methyl-N-propynyl(2-phenyl-1-methyl)ethylammonium hydrochloride L-deprenyl) to rats resulted in transfer of the radiolabelled methyl group to the Nepsilon-amino group of the endogenous lysine. The radiolabelled Nepsilon-monomethyllysine was urinary eliminated together with the other radiolabelled deprenyl metabolites, such as deprenyl-N-oxide and methamphetamine. The presence of Nepsilon-monomethyllysine has also been traced, and its concentrations were compared in the serum, liver and brain of rats subjected to L-deprenyl treatment. Methyl group transfer from the L-deprenyl to endogenous compounds; and the urinary elimination of their products may offer a vital way to eliminate or to decrease the degree of drug transmethylation to the lysine constituents of blood vessels' proteins.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Nε-Methyl-L-lysine hydrochloride, ≥98.0% (TLC)