MilliporeSigma
Oxidative DNA damage induced by a melatonin metabolite, 6-hydroxymelatonin, via a unique non-o-quinone type of redox cycle.
Biochemical pharmacology (2004-09-29)
Katsuhisa Sakano, Shinji Oikawa, Yusuke Hiraku, Shosuke Kawanishi
RÉSUMÉ
Melatonin, an indolic pineal hormone, is produced primarily at night in mammals and is important in controlling biological rhythms. Although melatonin is known to be effective as a free radical scavenger and has an anti-cancer effect, carcinogenic properties have also been reported. In relation to its carcinogenic potential, we have examined whether 6-hydroxymelatonin, a major melatonin metabolite, can induce DNA damage in the presence of metal ion using [32P]-5'-end-labeled DNA fragments obtained from genes relevant to human cancer. 6-Hydroxymelatonin induced site-specific DNA damage in the presence of Cu(II). Formamidopyrimidine-DNA glycosylase treatment induced cleavage sites mainly at G residues of the 5'-TG-3' sequence, whereas piperidine treatment induced cleavage sites at T mainly of 5'-TG-3'. Interestingly, 6-hydroxymelatonin strongly damaged G and C of the 5'-ACG-3' sequence complementary to codon 273 of the p53 gene. These results suggest that 6-hydroxymelatonin can cause double-base lesions. DNA damage was inhibited by both catalase and bathocuproine, Cu(I)-specific stabilizer, suggesting that reactive species derived from the reaction of H2O2 with Cu(I) participate in DNA damage. Cytochrome P450 reductase efficiently enhanced 6-hydroxymelatonin-induced oxidative DNA damage and oxygen consumption, suggesting the formation of redox cycle. It is noteworthy that 6-hydroxymelatonin can efficiently induce DNA damage via non-o-quinone type of redox cycle. Formation of 8-oxo-7,8-dihydro-2'-deoxyguanosine (8-oxodG), a characteristic oxidative DNA lesion, in calf thymus DNA was significantly increased by 6-hydroxymelatonin in the presence of Cu(II). Furthermore, 6-hydroxymelatonin significantly increased the formation of 8-oxodG in human leukemia cell line HL-60 but not in HP100, a hydrogen peroxide (H2O2)-resistant cell line derived from HL-60. The 6-hydroxymelatonin-induced 8-oxodG formation in HL-60 cells significantly decreased by the addition of bathocuproine or o-phenanthroline. Therefore, it is concluded that melatonin may exhibit carcinogenic potential through oxidative DNA damage by its metabolite.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
6-Hydroxymelatonin