MilliporeSigma
  • Polymeric drugs with prolonged sustained delivery of specific anti-aggregant agents for platelets: kinetic analysis of the release mechanism.
Polymeric drugs with prolonged sustained delivery of specific anti-aggregant agents for platelets: kinetic analysis of the release mechanism.
Journal of biomaterials science. Polymer edition (2004-08-21)
Alberto Gallardo, Gema Rodríguez, Mar Fernández, María Rosa Aguilar, Julio San Román
RÉSUMÉ
The in vitro aqueous behaviour of a metacryloyloxyethyl [2-(acetyloxy)-4-(trifluoromethyl)]benzoate (THEMA)/N,N'-dimethylacrylamide (DMA) copolymer with a THEMA molar content of 39% (labeled THDMA39) has been investigated. This composition has been selected to achieve a system able to keep both the non-water solubility during the release and the resorbability (and the water solubility) after the completion of the drug release. This copolymer exhibited, at pH 7.4, a constant release during several months, very interesting for a long term treatments required for the application of some cardiovascular devices. A kinetic model has been developed to explain the pseudo-zero-order kinetics of the release process. This model, which considers (from the aqueous studies) a linear increase with time of the amount of water present in the polymeric matrix, has been able to fit adequately the experimental data.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-(Trifluoromethyl)benzoic acid, 99%