MilliporeSigma
Suspected cases of bromocyclen poisoning.
Veterinary and human toxicology (1992-10-01)
M P Quick
RÉSUMÉ
This report summarizes the case histories and tabulates the analytical results from investigations of 8 incidents of suspected bromocyclen poisoning. Interpretation of the results and withdrawal of ALUGAN from the UK market are discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Bromocyclen, PESTANAL®, analytical standard