MilliporeSigma
Compatibility of the S-(3-nitro-2-pyridinesulfenyl) protecting group with DCC/HOBt coupling chemistry.
Peptide research (1992-09-01)
R Matsueda, S Higashida, F Albericio, D Andreu
RÉSUMÉ
Two recent reports on the partial lability of the 3-nitro-2-pyridinesulfenyl (Npys) thiol protecting group towards 1-hydroxy-benzotriazole (HOBt) have prompted a rechecking of the chemical behavior of this group. Using both soluble and polymer-bound forms of Cys(Npys) as test materials, the complete stability of this protection against HOBt has now been definitively established, and its compatibility with Boc-benzyl-based solid-phase synthesis strategies has been clearly confirmed by stability assays against a wide range of reagents, as well as by the successful synthesis of several Cys(Npys)-containing peptides.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Nitro-2-pyridinesulfenyl chloride, 95%