MilliporeSigma
The dimensional accuracy of rectangular acrylic resin specimens cured by three denture base processing methods.
The Journal of prosthetic dentistry (1992-06-01)
S Salim, S Sadamori, T Hamada
RÉSUMÉ
The dimensional accuracy of rectangular acrylic resin specimens was examined when they were processed by three methods: a conventional method, the SR-Ivocap system, and a microwave curing method. The dimensional accuracy was evaluated by the change of the distance vector V, which is calculated by means of measurements of the distances between fixed points on specimens. The specimen cured by the SR-Ivocap system exhibited less dimensional change (p less than 0.05) than those cured by the conventional and the microwave curing methods. The SR-Ivocap system might produce a more accurate denture base than the conventional and the microwave curing methods.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Poly(methyl methacrylate-co-ethylene glycol dimethacrylate), 50 μm particle size