MilliporeSigma
Genetic breeding of L-tyrosine producer from Brevibacterium lactofermentum.
Agricultural and biological chemistry (1990-03-01)
H Ito, S Sakurai, T Tanaka, K Sato, H Enei
RÉSUMÉ
A wild-type parent of Brevibacterium lactofermentum was converted into an L-Tyr producer by three steps of genetic breeding. First, acquirement of m-fluoro-D, L-phenylalanine resistance (1,000 microgram/ml) brought about MF1317 which produced 3.5 g/l of L-Tyr and a byproduct of 2.8 g/l of L-Phe. Second, increase in the drug resistance (5,000 microgram/ml) gave MF358 that produced 6.4 g/l of L-Tyr and a byproduct of 6.0 g/l of L-Phe. Third, an L-Phe auxotrophic mutant (FT-1) derived from MF358 accumulated 16 g/l of L-Tyr. In FT-1, L-Phe was not accumulated at all, but a small amount of anthranilate (0.4 g/l) was. A key enzyme in the biosynthesis of L-Tyr, 3-deoxy-D-arabino-heptulosonate-7-phosphate synthase, was free from synergistic feedback inhibition by L-Tyr and L-Phe in the producers, and so L-Tyr accumulation occurred independently of L-Phe concentration in the production medium.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
m-Fluoro-DL-phenylalanine