MilliporeSigma
A bi-symmetric square wave Zeeman modulator for nuclear quadrupole resonance.
Solid state nuclear magnetic resonance (1992-11-01)
D Mao, G L Petersen, P J Bray
RÉSUMÉ
A simple circuit has been designed to generate a bi-symmetric square wave Zeeman modulation for the detection of nuclear quadrupole resonance. The square waveform not only provides an optimum result among bi-symmetric modulation waveforms, but also allows the observation of the Zeeman perturbed NQR powder pattern without the need for an extra external magnetic field.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Calcium metaborate, technical, 39-44% B2O3 basis, 31-37% CaO basis, powder